Super Bonder® 499™ Thermal Cycling-Resistant Gel Adhesive, Loctite®, Henkel
Supplier: Loctite
135471
82002-818CS 338.09 USD
82002-818
Super Bonder® 499™ Thermal Cycling-Resistant Gel Adhesive, Loctite®, Henkel
Adhesives
This thixotropic high-viscosity, high-temperature gel formulation bonds all metals, and most plastics and rubber compounds.
  • High-viscosity
  • High-temperature
  • Fills gaps up to 0.025cm (0.01")
Adhesive is suitable for use with vertical or overhead surfaces and parts that require thermal cycling resistance.