Super Bonder® 430™ Instant Adhesive, Loctite®, Henkel
Supplier: Loctite
233978
82002-810CS 332.2 USD
82002-810
Super Bonder® 430™ Instant Adhesive, Loctite®, Henkel
Adhesives
Super Bonder® 430™ adhesive is a general industrial-grade cyanoacrylate adhesive.
  • Industrial-grade
  • Fills gaps up to 0.012cm (0.005")
Adhesive is for close-fitting metallic parts.
Certifications: Commercial item std. A-A-3097, Type I, Class 2.