Assure™ 425™ Surface-Curing Threadlocker, Loctite®, Henkel
Supplier: Loctite
135461
82002-808CS 335.53 USD
82002-808
Assure™ 425™ Surface-Curing Threadlocker, Loctite®, Henkel
Adhesives
This low-strength, fast-curing threadlocker was developed for locking fasteners on plastic or metal parts.
  • Low-strength
  • Fast-curing
  • Can be post-applied
Recommended for nylon, polyester, and other plastic surfaces. Store refrigerated.