Super Bonder® 420™ Instant Adhesive, Loctite®, Henkel
Supplier: Loctite
135455
82002-804CS 324.96 USD
82002-804
Super Bonder® 420™ Instant Adhesive, Loctite®, Henkel
Adhesives
Super Bonder® 420™ adhesive is designed to penetrate by capillary action between preassembled components.
  • Instant adhesive
  • Fills gaps up to 0.005cm (0.002")
Parts may be made of metal, plastic, or elastomer materials and must be close-fitting.
Certifications: Commercial item std. A-A-3097, Type II, Class 1.