PRISM® 411™ Clear Toughened Instant Adhesive, Loctite®, Henkel
Supplier: Loctite
135446
82002-796CS 362.71 USD
82002-796
PRISM® 411™ Clear Toughened Instant Adhesive, Loctite®, Henkel
Adhesives
High-viscosity, toughened ethyl adhesive bonds rubber, plastic, and metal substrates.
  • Tough adhesive
  • High-viscosity
  • Fills gaps up to 0.020cm (0.008")
Adhesive is ideal for applications where gap-filling is required. It is peel- and shock-resistant.