PRISM® 410™ Black Toughened Instant Adhesive, Loctite®, Henkel
Supplier: Loctite
135444
82002-794CS 365.37 USD
82002-794
PRISM® 410™ Black Toughened Instant Adhesive, Loctite®, Henkel
Adhesives
This black, toughened ethyl adhesive bonds rubbers, metals, and plastics for use in difficult environments.
  • Tough adhesive
  • For use in difficult environments
  • Fills gaps up to 0.020cm (0.008")
Adhesive is peel- and shock-resistant.