PRISM® 408™ Low Odor/Low Bloom Instant Adhesive, Loctite®, Henkel
Supplier: Loctite
135441
82002-792CS 7673.61 USD
82002-792
PRISM® 408™ Low Odor/Low Bloom Instant Adhesive, Loctite®, Henkel
Adhesives
This low-viscosity adhesive instantly bonds components with minimal fumes or odors and has non-blooming characteristics.
  • Instantly bonds components
  • Minimal fumes or odors
  • Non-blooming characteristics
Adhesive is designed to penetrate between close-fitting or preassembled parts with gaps up to 0.005cm (0.002").