PRISM® 406™ Instant Adhesive, Surface Insenstive, Loctite®, Henkel
Supplier: Loctite
135436
82002-790CS 775.65 USD
82002-790
PRISM® 406™ Instant Adhesive, Surface Insenstive, Loctite®, Henkel
Adhesives
This low-viscosity adhesive is excellent for almost all surfaces.
  • 3,200 psi tensile strength
  • Fills gaps up to 0.010 cm (0.004")
  • Wicking grade viscosity
  • Fast curing
Ideal for difficult-to-bond plastic and rubber compounds.