Loctite® 3311 Light Cure Adhesive, Henkel
Supplier: Loctite
88189
82002-774CS 1113.74 USD
82002-774
Loctite® 3311 Light Cure Adhesive, Henkel
Adhesives
Ultraviolet light cure medical device adhesive that comes in a syringe.
  • Pale yellow in color
  • Great for bonding thermoplastics and plasticized PVC
  • Comes in a syringe