Loctite® 3301™ Light Cure Adhesive, Henkel
Supplier: Loctite
475362
82002-772CS 571.73 USD
82002-772
Loctite® 3301™ Light Cure Adhesive, Henkel
Adhesives
Loctite® 3301™ one-part acrylate urethane light cure adhesive is pale yellow in color and has a vapor pressure greater than 5 millimeters Hg at 20 degrees C.
  • Excellent adhesion to a wide variety of substrates including glass, many plastics and most metals
  • Suitable for use in the assembly of disposable medical devices
  • UV/ visible light cure: Up to 80s
  • Free from fluorescent
The liquid adhesive has a mild odor and comes in a 25-milliliter syringe.
Loctite 3301 is primarily designed for bonding rigid or flexible PVC to polycarbonate enabling easy assembly of components with close fitting tolerances (i.e. joining polycarbonate to flexible PVC tubing).