PRISM® 4011™ Low Viscosity Instant Adhesive, Loctite®, Henkel
Supplier: Loctite
142059
82002-768CS 910.81 USD
82002-768
PRISM® 4011™ Low Viscosity Instant Adhesive, Loctite®, Henkel
Adhesives
LOCTITE® 4011™ Prism® low viscosity is a one part, room temperature cure, surface insensitive, instant adhesive.
  • Designed for the assembly of difficult-to-bond materials which require uniform stress distribution
  • Suited for bonding a wide range of materials, including metals, plastics, and elastomers
Viscosity Temperature: 25°C (77°F).
Certifications: ISO-10993 Biological Tested for medical device use