Monoject™ Allergy Syringes with Permanently Attached Needles, in Trays, Covidien
Supplier: Cardinal Health
8881500501
82001-194CS 676.82 USD
82001-194
Monoject™ Allergy Syringes with Permanently Attached Needles, in Trays, Covidien
Syringes Syringes, Medical
Each tray contains 25 sterile, 28-gauge allergy syringes with an ultra-comfort bevel for injection
The syringes feature a flat plunger tip and graduations at 0.01cc. Latex free.