Agar for bacteriology

Supplier: VWR

Synonyms: Agar Agar

J637-1KG J637-500G J637-2.5KG J637-10KG
97064-332CS 1502.48 USD
97064-332 97064-336 97064-334 80760-978
Agar for bacteriology
Agar

A solidifying agent used in bacteriology. Agar consists of unbranched polysaccharides of galactose subunits derived from algae or seaweed.


  • Agar for the growth of bacteria and fungi
  • Typically used at concentrations of 1-2% in solid media preparation

Melting Pt: 90
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
Merck Index: 13,00184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR