L-Alanine

Supplier: VWR

Synonyms: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine

BRGT106-5KG
80760-316EA 250.96 USD
80760-316
L-Alanine
L-Alanine
Formula: C₃H₇NO₂
MW: 89.09 g/mol
MDL Number: MFCD00064410
CAS Number: 56-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR