Disodium hydrogen phosphate, anhydrous ≥98.0%, Reagent Grade
Supplier: Avantor Performance Materials
Synonyms:
sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic
MS0876-25KG
80731-078EA
1406.52
USD
80731-078
Disodium hydrogen phosphate, anhydrous ≥98.0%, Reagent Grade
di-Sodium hydrogen phosphate
Formula:
Na₂HPO₄ MW: 141.96 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003496 CAS Number: 7558-79-4 |
Specification Test Results
Assay | Min. 98.0% |
Appearance | White Crystalline Powder |
Solubility in water (1 g/10 ml) | Clear and colorless |
pH (1%) | 8.7 - 9.6 |
Heavy metals (as Pb) | Max. 10 ppm |
Insoluble Matter | Max. 0.2% |
Loss on Drying | Max. 5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...