Hydrochloric acid 36.5 - 38.0% ACS

Supplier: Anachemia
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

5100-4G250C 5100H-4H250C 46414-461
CA11020-880CS 631.51 USD
CA11020-880 CA11020-884 80722-152
Hydrochloric acid 36.5 - 38.0% ACS
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Assay (as HCl) 36.5 - 38.0%
Appearance Passes Test
Colour (APHA) 10 max.
Residue after ignition 5 ppm max.
Bromide (Br) 0.005% max.
Sulfate (SO4) 1 ppm max.
Sulfite (SO3) 1 ppm max.
Extractable organic substances Passes Test (about 5 ppm)
Free Chlorine (Cl) 1 ppm max.
Ammonium (NH4) 3 ppm max.
Arsenic (As) 0.01 ppm max.
Heavy metals (as Pb) 1 ppm max.
Iron (Fe) 0.2 ppm max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR