Sodium borohydride, crystalline powder ACS

Supplier: SPECTRUM LABORATORY PRODS LO
Danger

S1187-10
80701-854EA 1484.44 USD
80701-854
Sodium borohydride, crystalline powder ACS
Sodium borohydride
Formula: NaBH₄
MDL Number: MFCD00003518
CAS Number: 16940-66-2
UN: 1426
ADR: 4.3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR