INSWOOL®-HP Blanket, HARBISON-WALKER
Supplier: HARBISON-WALKER
69998
80701-814EA 416.59 USD
80701-814
INSWOOL®-HP Blanket, HARBISON-WALKER
Insulation
Refractory ceramic fiber (RCF) blanket rated at 2,300 degrees F.
  • Flexible and lightweight
  • Highly insulating and thermal shock resistant
  • Not suitable for environments with high gas velocities or particulates
It has excellent strength, both hot and cold. It remains in place on the furnace anchors even at high temperatures and can resist damage even when subjected to normal mistreatment in shipment and handling.