Sodium carbonate light

Warning

16141020
80701-656EA 103.53 USD
80701-656
Sodium carbonate light
Sodium carbonate

Soda ash (sodium carbonate) is a basic component found in fire assay flux. In either light or dense format it reacts with silica based minerals to form fusible sodium silicates.

Formula: Na₂CO₃
MDL Number: MFCD00003494
CAS Number: 497-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR