HgX® Mercury Decontaminant Powder, Action Technologies
Supplier: ACTON TECHNOLOGIES
HGX-10LB
80701-498EA 290.63 USD
80701-498
HgX® Mercury Decontaminant Powder, Action Technologies
Cleaning Agents Decontaminants
Mercury decontaminant powder reacts rapidly and forming a film over the finely divided, nearly invisible beads of mercury and reacting to produce a non-vaporizing sulfide. It is used for the reduction of toxic mercury vapor.
It is not harmful. None of its components are classified as toxic. It leaves no odor. It has no allergic reactions even after continuous use under various conditions.