Diatomaceous earth (DE), filter white flux (calcined), Celatom® FW-18
|
|
Synonyms:
Kieselguhr
80701-462EA
76.24
USD
80701-462
Diatomaceous earth (DE), filter white flux (calcined), Celatom® FW-18
Diatomaceous earth (DE), filter white flux (calcined)
Filter aid with wide range of applications
Specification Test Results
Color | White to Pinkish |
Sieve Analysis (Tyler) % + 150 Mesh (> 105 microns) | 7.6 |
Median Particle Diameter (microns) | 31.0 |
pH (10% Slurry) | 9.5 |
Density (dry bulk) g/l | 224 |
Density (wet bulk) g/l | 336 |
Permeability (millidarcy) | 1900 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...