Potassium nitrate (nitre- KNO₃)

Warning

16153637
80701-370EA 309.89 USD
80701-370
Potassium nitrate (nitre- KNO₃)
Potassium nitrate
Formula: KNO₃
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
UN: 1486
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR