Aluminium oxide ≥97%, GREENCAST® 97 PLUS

Supplier: HARBISON-WALKER

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

59338
80700-882EA 483.3 USD
80700-882
Aluminium oxide ≥97%, GREENCAST® 97 PLUS
Aluminium oxide

GREENCAST® 97 PLUS is a 97% alumina dense castable for temperatures up to 3400 °F (1870 °C). It is based on very high purity ingredients, low in silica, iron oxide and alkalies.

Formula: Al₂O₃
MW: 101.96 g/mol
MDL Number: MFCD00003424
CAS Number: 1344-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR