Sodium hydrogen carbonate USP

Supplier: UNIVAR USA INC

Synonyms: Sodium bicarbonate

89362
80700-802EA 200.9 USD
80700-802
Sodium hydrogen carbonate USP
Sodium hydrogen carbonate
Formula: NaHCO₃
MW: 84.01 g/mol
MDL Number: MFCD00003528
CAS Number: 144-55-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR