Sodium hydroxide 50%

Supplier: SIERRA CHEMICAL COMPANY
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

261491-700LB
80700-750EA 917.81 USD
80700-750
Sodium hydroxide 50%
Sodium hydroxide
Formula: NaOH
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR