SP Bel-Art Acid/Solvent Bottle Carriers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
169590000
80700-744CS 767.58 USD
80700-744
SP Bel-Art Acid/Solvent Bottle Carriers, Bel-Art Products, a part of SP
Carriers Bottle Carriers/Containers
Polyethylene carriers with epoxy-coated, heavy-gauge steel wire bails feature wide ribs on both the sides and the bottom to support and cushion bottles.
  • Truncated style lid permits pouring without the removal of the lid
  • Designed to hold standard 2.5 L (5 pt.) US reagent bottles