Crucible Pouring Tongs, Legend
Supplier: LEGEND, INC
19224-50
80700-696EA 208.82 USD
80700-696
Crucible Pouring Tongs, Legend
Tongs
Crucible tongs are made of steel for durability.
  • Increase safety by distancing the user from the oven or furnace
'Hairpin' style pouring tong which clamps the crucible into the ring. Crucible will not fall out of tong when clamped.