Flour All Purpose
8843
80700-684EA 89.35 USD
80700-684
Flour All Purpose
Flour
Flour is a commonly used reducing agent in Fire Assay flux. It is used to reduce litharge to molten lead for the collection of precious metal in the sample.