Sodium nitrate

16142300
80700-093EA 142.32 USD
80700-093
Sodium nitrate
Sodium nitrate
Formula: NaNO₃
MW: 84.99 g/mol
MDL Number: MFCD00011119
CAS Number: 7631-99-4
UN: 1498
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR