Sand, quartz, Premier Silica 6/9

Supplier: P.W. GILLIBRAND CO. INC.

12521
80700-084EA 2495 USD
80700-084
Sand, quartz, Premier Silica 6/9
Sand, quartz

Silica sand provided from high quality natural deposits.


The sand is quarried or mined and then washed, dried and screened to produce the optimum particle size distribution for the intended end use. Pulverized silica (also known as silica flour) is a fine powder form of silica sand used to manufacture fiberglass insulation, ceramic products including tableware, sanitary ware, tile, caulks, paints, plastics and many others.

Formula: SiO₂
MDL Number: MFCD02100519
CAS Number: 14808-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR