Lead(II) oxide (Litharge), powder gold- and silver-free

Danger

MR111-25KG
80700-079EA 485.62 USD
80700-079
Lead(II) oxide (Litharge), powder gold- and silver-free
Lead(II) oxide (Litharge)

Litharge is a unique type of lead oxide used in the Fire Assay analysis of precious metals. It is produced from highly refined lead and offers a very low level of precious metal impurities.

Formula: PbO
Boiling Pt: 1470 °C (1013 hPa)
Melting Pt: 888 °C
Density: 9.5 g/cm³ (20 °C)
MDL Number: MFCD00011164
CAS Number: 1317-36-8
EINECS: 215-267-0
UN: 2291
ADR: 6.1,III
Merck Index: 13,05431

Order Now

Specification Test Results

Appearance granularity Granular
Appearance colour Yellow to orange


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR