RNase A, Protease-Free, Highly Purified, Bovine Pancreas, MilliporeSigma
Supplier: MilliporeSigma
556746-50KU
556746-10KU
80602-646EA
633.29
USD
80602-646
80602-644
RNase A, Protease-Free, Highly Purified, Bovine Pancreas, MilliporeSigma
Media for Microbiology
Dehydrated Media for Microbiology
Ribonuclease A, Protease-Free is a chromatographically purified, pyrimidine-specific endoribonuclease that acts on single-stranded RNA
For selective degradation of RNA.
One unit is defined as the amount of enzyme that will catalyze the hydrolysis of RNA to yield a first-order velocity constant equal to 1 at 25°C, pH 5.0.