Glycogen (from mussels) for molecular biology, Calbiochem®

Supplier: MilliporeSigma

361507-1ML
80601-860EA 488.32 USD
80601-860
Glycogen (from mussels) for molecular biology, Calbiochem®
Glycogen
Formula: C₂₄H₄₂O₂₁
MW: 666.579 g/mol
MDL Number: MFCD00081547
CAS Number: 9005-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR