Octaethylene glycol monododecylether (C12E8) solution 10%

Supplier: MilliporeSigma

Synonyms: 3,6,9,12,15,18,21,24-Octaoxahexatriacontan-1-ol, Octaethylene glycol monododecyl ether, Octaethylene glycol monolauryl ether

205532-1SET
80600-990EA 448.17 USD
80600-990
Octaethylene glycol monododecylether (C12E8) solution 10%
Octaethylene glycol monododecylether (C12E8)
Formula: C₂₈H₅₈O₉
MW: 538.754 g/mol
MDL Number: MFCD00043234
CAS Number: 3055-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR