TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) non-ionic detergent, Calbiochem®

Supplier: MilliporeSigma
Danger

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

648462-3KG 648462-1KG
80512-694EA 374.2 USD
80512-694 80057-664
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) non-ionic detergent, Calbiochem®
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)

Non-ionic detergent and emulsifier.

Melting Pt: 6
Flash Pt: 251
Storage Temperature: Refrigerator
MDL Number: MFCD00132505
CAS Number: 9002-93-1
UN: 3082
ADR: 9 III
Merck Index: 13,06793

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR