2,2-Diphenyl-1-picrylhydrazyl (free radical) ≥90%, Calbiochem®

Supplier: MilliporeSigma
Danger

Synonyms: 2,2-Diphenyl-1-(2,4,6-trinitrophenyl)hydrazyl free radical, DPPH free radical

Packaged under inert gas.

300267-50MG
80512-444EA 125.32 USD
80512-444
2,2-Diphenyl-1-picrylhydrazyl (free radical) ≥90%, Calbiochem®
2,2-Diphenyl-1-picrylhydrazyl (free radical)

DPPH, Free Radical, is a cell-permeable, stable free radical that acts as a hydrogen radical scavenger.


  • Useful in inducing free-radical injury to tissues
  • Screening tool for detecting free radical scavenging activity of antioxidants
  • Absorption max: ~517 nm
  • Soluble in DMF or EtOH


Special Instructions: Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. Unstable in aqueous solutions.

Formula: C₁₈H₁₂N₅O₆
MW: 394.32 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00007231
CAS Number: 1898-66-4
EINECS: 217-591-8

Order Now

Specification Test Results

Purity by Elememtal Analysis ≥90%
Form Black solid
Toxicity Harmful
Storage -20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR