KOD XL DNA Polymerase, MilliporeSigma
Supplier: MilliporeSigma
71087-4
80511-390EA 2912.12 USD
80511-390
KOD XL DNA Polymerase, MilliporeSigma
Nucleic Acid Reagents End-point PCR Enzymes and Kits
KOD XL DNA Polymerase is an optimized blend of KOD DNA Polymerase and a mutant form of KOD that is deficient in 3′→5′ exonuclease activity.
  • Ideal for amplification of large DNA fragments from purified DNA or crude samples
  • Amplifies DNA templates up to 30 kbp
  • Successfully amplifies GC-rich sequences
  • Efficiently incorporates derivatized dNTPs
The enzyme mixture is designed for reliable amplification of long, complex targets with robust yield and high accuracy. It can also be used for incorporation of derivatized dNTPs in PCR amplicons. It generates a mixture of PCR products with blunt and 3′-nucleotide overhangs, suitable for cloning with various vector kits.