GeneJuice® Transfection Reagent, Novagen®, MilliporeSigma
Supplier: MilliporeSigma
GeneJuice® Transfection Reagent is a proprietary formulation optimized for maximal transfection efficiency, ease of use, and minimal cytotoxicity. This transfection reagent is a superior alternative to a wide variety of other techniques including calcium phosphate coprecipitation, electroporation, microinjection, biolistic particle delivery, and complex formation with DEAE-dextran.
GeneJuice Transfection Reagent provides excellent performance in both stable and transient transfection of eukaryotic cells and is ideal for use with the Novagen pTriEx™ expression vectors.
The 1 ml size provides enough reagent to perform up to 500 transfections in standard 35 mm plates. The reagent is also available in an introductory 0.3 ml size, a convenient, new 5 × 1 ml size, and a 10 ml size. GeneJuice Transfection Reagent is supplied as a ready-to- use sterile solution.