1,10-Phenanthroline monohydrate ≥99%, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1,10-Phenanthroline hydrate

Packaged under inert gas.

516705-500MG
80511-070EA 96.81 USD
80511-070
1,10-Phenanthroline monohydrate ≥99%, Sigma-Aldrich®
1,10-Phenanthroline monohydrate

A metal chelating agent that prevents the induction of chromosomal aberrations in streptozotocin-treated cells. Acts as a metalloprotease inhibitor. Inhibits glioma invasion in vitro.

Formula: C₁₂H₈N₂·1H₂O
MW: 198.22 g/mol
Melting Pt: 93…94 °C
Density: 1.259 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149973
CAS Number: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1,III
Merck Index: 13,07291
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba346b9f67fdda9afd4d48b893e6318564d604c&vid=2d08e04ee9525293a747b55cdcff8f66a6c4715a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Apperance Off-white solid
Purity by titration ≥99%
Solubility DMSO (100 mg/mL), ethanol (25 mg/mL), H2O (3 mg/mL)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR