PIPPS (1,4-Piperazinedipropanesulfonic acid)

Supplier: MilliporeSigma

Synonyms: Piperazine-N,N'-bis(3-propanesulphonic acid), Piperazine-N,N'-bis(3-propanesulfonic acid), Piperazine-1,4-bis(propanesulphonic acid), Piperazine-1,4-bis(propanesulfonic acid)

528315-100GM
80511-042EA 429.55 USD
80511-042
PIPPS (1,4-Piperazinedipropanesulfonic acid)
PIPPS (1,4-Piperazinedipropanesulfonic acid)
Formula: C₁₀H₂₂N₂O₆S₂
MW: 130.093 g/mol
MDL Number: MFCD00274122
CAS Number: 5625-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR