Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.0% (by titrimetric analysis), Molecular Biology Grade, Calbiochem®

Supplier: MilliporeSigma
Warning

648310-500GM
80503-542EA 244.92 USD
80503-542 80503-540
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.0% (by titrimetric analysis), Molecular Biology Grade, Calbiochem®
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)

TRIS base is useful in the pH range of 7.0-9.0. Has a pKa of 8.1 at 25°C.

Formula: NH₂C(CH₂OH)₃
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 169…172 °C
Density: 1.328 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

pKa 8.30 at 20°C
Purity ≥99% by titration test (perchloric acid titration)
Absorbance (260/280 nm) ≤0.03/≤0.02
Heavy metals ≤0.0005%
RNases, DNases, proteases none detected
Storage 15 to 30 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR