Tween® 20 (Polysorbate), Molecular Biology Grade detergent, Calbiochem®, Millipore®

Supplier: MilliporeSigma

Synonyms: Polyoxyethylene sorbitan monolaurate

655204-100ML
80503-492EA 88.31 USD
80503-492
Tween® 20 (Polysorbate), Molecular Biology Grade detergent, Calbiochem®, Millipore®
Tween® 20 (Polysorbate)

TWEEN® 20 Detergent is a molecular biology grade, pale yellow liquid that offeres a specific gravity of 1.100 to 1.110 (at 20°C)

Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR