4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL)

Supplier: MilliporeSigma

Synonyms: 2,2,6,6-Tetramethyl-4-hydroxy-1-piperidinyloxy radical, 4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, Tempol

581500-500MG
80503-466EA 114.99 USD
80503-466
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL)
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-Hydroxy-TEMPO, TEMPOL, free radical)
Formula: C₉H₁₈NO₂
MW: 172.25 g/mol
MDL Number: MFCD00006478
CAS Number: 2226-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR