tri-Sodium citrate dihydrate, Molecular Biology Grade, Calbiochem®, Millipore®
Supplier: MilliporeSigma
Synonyms:
Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Trisodium citrate, Sodium Citrate, Dihydrate, Sodium citrate dihydrate, Citric acid tridosium salt dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate
M-Clarity™ quality level = MQ100
567446-1KG
567446-100GM
567446-5KG
80503-404EA
107.04
USD
80503-404
80503-402
80503-406
tri-Sodium citrate dihydrate, Molecular Biology Grade, Calbiochem®, Millipore®
tri-Sodium citrate dihydrate
Formula:
Na₃C₆H₅O₇·2H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150031 CAS Number: 6132-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...