Sodium dodecyl sulfate (SDS) ≥99%, Molecular Biology Grade, Calbiochem®, Millipore®

Supplier: MilliporeSigma
Danger

428023-500GM
80503-356EA 526.84 USD
80503-356
Sodium dodecyl sulfate (SDS) ≥99%, Molecular Biology Grade, Calbiochem®, Millipore®
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99%
DNases, proteases, RNases none detected
Heavy metals ≤0.0005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR