Polyethylene glycol 6,000, Molecular Biology Grade, Calbiochem®

Supplier: MilliporeSigma

Synonyms: PEG 6,000, PEG 6000

528877-1KG
80503-176EA 312.29 USD
80503-176
Polyethylene glycol 6,000, Molecular Biology Grade, Calbiochem®
Polyethylene glycol 6,000

Polyethylene glycol is used as stationary phase in gas chromatography.

Formula: H(OCH₂CH₂)nOH
MW: 6000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01779614
CAS Number: 25322-68-3

Order Now

Specification Test Results

Form White solid
Storage 15 to 30 °C
absorbance at 260 nm ≤0.2
absorbance at 280 nm ≤0.1
Heavy metals (as Pb) ≤0.0005%
DNase, proteases, RNase None detected
Absorbance (260 nm) ≤0.2
Absorbance (280 nm) ≤0.1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR