Magnesium chloride hexahydrate, Molecular Biology Grade, Calbiochem®

Supplier: MilliporeSigma

442611-500GM
80503-016EA 85.51 USD
80503-016
Magnesium chloride hexahydrate, Molecular Biology Grade, Calbiochem®
Magnesium chloride hexahydrate

Ultrapure Solutions® 10704S1G15 SemiConductor Grade NMP (1-Methyl-2-Pyrrolidinone).

Formula: MgCl₂·6H₂O
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Purity ≥99% by complexometry
Contaminants DNases, proteases, RNases: none detected
Heavy metals ≤0.0005% (as Pb)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR