MOPS, Free Acid, Molecular Biology Grade, EMD Millipore
Supplier: MilliporeSigma
475922-100GM
80503-004EA
133.87
USD
80503-004
MOPS, Free Acid, Molecular Biology Grade, EMD Millipore
Buffers
Electrophoresis Buffers
Running Buffers
MOPS, free acid is a zwitterionic buffer useful in the pH range of 6.5 to 7.9. Has a pKa of 7.20 at 25 °C.
- Purity: ≥99% (Potentiometry)
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)