Kifunensine (from Kitasatosporia kifunense) ≥98% (by TLC), Calbiochem®

Supplier: MilliporeSigma

422500-1MG
80502-850EA 399.74 USD
80502-850
Kifunensine (from Kitasatosporia kifunense) ≥98% (by TLC), Calbiochem®
Kifunensine

A potent alkaloid inhibitor of mannosidase I. Does not affect mannosidase II and the endoplasmic reticulum α-mannosidase.

Formula: C₈H₁₂N₂O₆
MW: 232.192 g/mol
CAS Number: 109944-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR