Glycine ≥99%, Molecular Biology Grade, Calbiochem®

Supplier: MilliporeSigma

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

357002-1KG
80502-614EA 143.59 USD
80502-614
Glycine ≥99%, Molecular Biology Grade, Calbiochem®
Glycine

A non-essential amino acid. It can be used in the preparation of biological buffers.


  • Absorbance: ≤0.01 at 260 nm and 280 nm

Formula: NH₂CH₂COOH
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form White solid
Purity ≥99% (by titration)
Contaminants DNases, proteases, RNases: none detected.
Toxicity Standard Handling
Storage +15 °C - +30 °C
Solubility H2O (200 mg/ml)
Stability Following reconstitution, store at room temperature (20°C). Stock solutions are stable for up to 6 months at room temperature.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR